5-Chloropyrazine-2-carboxylic acid 95%

Supplier: Thermo Scientific
Warning

H32596.03
H32596.03EA 195 EUR
H32596.03
5-Chloropyrazine-2-carboxylic acid 95%
5-Chloropyrazine-2-carboxylic acid

Purity: 95% CAS No.: 36070-80-1 Molecular Formula: C5H3ClN2O2 Formula Weight: 158.54 Physical Form: Solid Melting Point: 150-160° MDL No.: MFCD09033269

Formula: C5H3ClN2O2
MW: 158,54 g/mol
Melting Pt: 150…160 °C
Storage Temperature: Ambient
MDL Number: MFCD09033269
CAS Number: 36070-80-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR