2-Methylhexanoyl chloride 97%
Supplier: Thermo Fisher Scientific
|
Danger
|
H32829.09
H32829.03
H32829.09EA
290
EUR
H32829.09
H32829.03
2-Methylhexanoyl chloride 97%
2-Methylhexanoyl chloride
Purity: 97%
CAS No.: 41693-47-4
Molecular Formula: C7H13ClO
Formula Weight: 148.63
Flash Point: 55°(131°F)
Refractive Index: 1.4290
Boiling Point: 163°
UN No.: UN2920
MDL No.: MFCD01318644
Formula:
C₇H₁₃ClO Boiling Pt: 163 °C Density: 0,957 Flash Pt: 55 °C (131 °F) Storage Temperature: Ambient |
MDL Number:
MFCD01318644 CAS Number: 41693-47-4 ADR: 8,II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...