4-Acetylbenzenesulphonamide 97%

Supplier: Thermo Fisher Scientific
Warning

H32901.03 H32901.06
H32901.03EA 62 EUR
H32901.03 H32901.06
4-Acetylbenzenesulphonamide 97%
4-Acetylbenzenesulphonamide

Purity: 97% CAS No.: 1565-17-9 Molecular Formula: C8H9NO3S Formula Weight: 199.23 Melting Point: 177-179° MDL No.: MFCD00792524

Formula: C₈H₉NO₃S
Melting Pt: 177…179 °C
Storage Temperature: Ambient
MDL Number: MFCD00792524
CAS Number: 1565-17-9
EINECS: 216-363-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR