3-(2-Benzothiazolyl)aniline 96%

Supplier: Thermo Scientific
Danger

H33103.MD H33103.03
H33103.MDEA 78.6 EUR
H33103.MD H33103.03
3-(2-Benzothiazolyl)aniline 96%
3-(2-Benzothiazolyl)aniline

Purity: 96% CAS No.: 41230-21-1 Molecular Formula: C13H10N2S Formula Weight: 226.30 Melting Point: 135-137° MDL No.: MFCD00579111

Formula: C₁₃H₁₀N₂S
MW: 226,30 g/mol
Melting Pt: 135…137 °C
Storage Temperature: Ambient
MDL Number: MFCD00579111
CAS Number: 41230-21-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR