5-(4-tert-Butylphenyl)-1,3,4-oxadiazole-2-thiol 96%

Supplier: Thermo Fisher Scientific
Warning

H33113.MD H33113.03
H33113.MDEA 0 EUR
H33113.MD H33113.03
5-(4-tert-Butylphenyl)-1,3,4-oxadiazole-2-thiol 96%
5-(4-tert-Butylphenyl)-1,3,4-oxadiazole-2-thiol

Purity: 96% CAS No.: 306936-90-3 Molecular Formula: C12H14N2OS Formula Weight: 234.32 Melting Point: 157-158° MDL No.: MFCD00500939

Formula: C₁₂H₁₄N₂OS
MW: 234.32 g/mol
Melting Pt: 157…158 °C
Storage Temperature: Ambient
MDL Number: MFCD00500939
CAS Number: 306936-90-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR