4-(2-Methoxyphenoxy)benzenesulphonyl chloride 96%
Supplier: Thermo Scientific
|
Danger
|
H33123.MD
H33123.03
H33123.06
H33123.MDEA
39.6
EUR
H33123.MD
H33123.03
H33123.06
4-(2-Methoxyphenoxy)benzenesulphonyl chloride 96%
4-(2-Methoxyphenoxy)benzenesulphonyl chloride
Purity: 96%
CAS No.: 690632-30-5
Molecular Formula: C13H11ClO4S
Formula Weight: 298.75
UN No.: UN3261
MDL No.: MFCD01631898
Formula:
C₁₃H₁₁ClO₄S MW: 298,75 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD01631898 CAS Number: 690632-30-5 ADR: 8,II |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...