3-(2-Methoxy-1-naphthyl)propionic acid 96%

Supplier: Thermo Fisher Scientific
Warning

H33173.MD H33173.03 H33173.06
H33173.MDEA 0 EUR
H33173.MD H33173.03 H33173.06
3-(2-Methoxy-1-naphthyl)propionic acid 96%
3-(2-Methoxy-1-naphthyl)propionic acid

Purity: 96% CAS No.: 34225-11-1 Molecular Formula: C14H14O3 Formula Weight: 230.26 Melting Point: 130-134° MDL No.: MFCD06823841

Formula: C₁₄H₁₄O₃
Melting Pt: 130…134 °C
Storage Temperature: Ambient
MDL Number: MFCD06823841
CAS Number: 34225-11-1

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR