4-Chloro-5-(4-methoxyphenyl)thieno[2,3-d]pyrimidine 96%
Supplier: Thermo Scientific
|
Warning
|
H33236.03
H33236.06
H33236.03EA
68.4
EUR
H33236.03
H33236.06
4-Chloro-5-(4-methoxyphenyl)thieno[2,3-d]pyrimidine 96%
4-Chloro-5-(4-methoxyphenyl)thieno[2,3-d]pyrimidine
Purity: 96%
CAS No.: 312584-53-5
Molecular Formula: C13H9ClN2OS
Formula Weight: 276.75
Melting Point: 166-168°
MDL No.: MFCD01831011
Formula:
C₁₃H₉ClN₂OS MW: 276,75 g/mol Melting Pt: 166…168 °C Storage Temperature: Ambient |
MDL Number:
MFCD01831011 CAS Number: 312584-53-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...