Methyl-1-benzylpyrrolidine-3-carboxylate ≥97%
Supplier: Thermo Scientific
Synonyms:
Methyl 1-benzylpyrrolidine-3-carboxylate
H33341.03
H33341.06
H33341.03EA
0
EUR
H33341.03
H33341.06
Methyl-1-benzylpyrrolidine-3-carboxylate ≥97%
Methyl-1-benzylpyrrolidine-3-carboxylate
Purity: 97%
CAS No.: 17012-21-4
Molecular Formula: C13H17NO2
Formula Weight: 219.28
Refractive Index: 1.5211
Boiling Point: 98°/0.2mm
MDL No.: MFCD04038678
Formula:
C₁₃H₁₇NO₂ MW: 219,28 g/mol |
MDL Number:
MFCD04038678 CAS Number: 17012-21-4 |
Learn more
![](/m-ie.vwr.com/en_IE/images/about_vwr_275x315.jpg)
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...