1-(3-Fluorophenyl)pyrrole 98%
Supplier: Thermo Fisher Scientific
H33348.03
H33348.06
H33348.03EA
0
EUR
H33348.03
H33348.06
1-(3-Fluorophenyl)pyrrole 98%
1-(3-Fluorophenyl)pyrrole
Purity: 98%
CAS No.: 53871-27-5
Molecular Formula: C10H8FN
Formula Weight: 161.18
Refractive Index: 1.5780
Boiling Point: 54°/0.15mm
MDL No.: MFCD02665249
Formula:
C₁₀H₈FN Boiling Pt: 54 °C (0,15 mmHg) Storage Temperature: Ambient |
MDL Number:
MFCD02665249 CAS Number: 53871-27-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...