2-Fluoro-4-methylsulphonylphenol 97%

Supplier: Thermo Fisher Scientific
Warning

H33358.MD H33358.03
H33358.MDEA 0 EUR
H33358.MD H33358.03
2-Fluoro-4-methylsulphonylphenol 97%
2-Fluoro-4-methylsulphonylphenol

Purity: 97% CAS No.: 398456-87-6 Molecular Formula: C7H7FO3S Formula Weight: 190.19 Melting Point: 92-94° MDL No.: MFCD09264549

Formula: C₇H₇FO₃S
MW: 190.19 g/mol
Melting Pt: 92…94 °C
Storage Temperature: Ambient
MDL Number: MFCD09264549
CAS Number: 398456-87-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR