4-(2-Methylphenoxy)benzenesulphonyl chloride 96%

Supplier: Thermo Fisher Scientific
Danger

H33556.03 H33556.06
H33556.03EA 0 EUR
H33556.03 H33556.06
4-(2-Methylphenoxy)benzenesulphonyl chloride 96%
4-(2-Methylphenoxy)benzenesulphonyl chloride

Purity: 96% CAS No.: 610277-83-3 Molecular Formula: C13H11ClO3S Formula Weight: 282.74 Melting Point: 81-85° UN No.: UN3261 MDL No.: MFCD01631900

Formula: C₁₃H₁₁ClO₃S
Melting Pt: 81…85 °C
Storage Temperature: Ambient
MDL Number: MFCD01631900
CAS Number: 610277-83-3
ADR: 8,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR