3-Amino-5-(4-chlorophenyl)-1,2,4-thiadiazole 96%

Supplier: Thermo Scientific
Danger

H33674.MD H33674.03
H33674.MDEA 37.5 EUR
H33674.MD H33674.03
3-Amino-5-(4-chlorophenyl)-1,2,4-thiadiazole 96%
3-Amino-5-(4-chlorophenyl)-1,2,4-thiadiazole

Purity: 96% CAS No.: 89894-30-4 Molecular Formula: C8H6ClN3S Formula Weight: 211.67 MDL No.: MFCD05865131

Formula: C₈H₆ClN₃S
MW: 211,67 g/mol
Storage Temperature: Ambient
MDL Number: MFCD05865131
CAS Number: 89894-30-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR