Ethyl-3-cyclopropyl-3-oxopropanoate 96%

Supplier: Thermo Scientific
Warning

Synonyms: Ethyl 3-cyclopropyl-3-oxopropanoate

H33756.03 H33756.06
H33756.03EA 34 EUR
H33756.03 H33756.06
Ethyl-3-cyclopropyl-3-oxopropanoate 96%
Ethyl-3-cyclopropyl-3-oxopropanoate
Formula: C₈H₁₂O₃
MW: 156,18 g/mol
Boiling Pt: 73 °C (1 mmHg)
Storage Temperature: Ambient
MDL Number: MFCD00184570
CAS Number: 24922-02-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR