Methyl-2-(trifluoromethyl)pyridine-5-carboxylate 97%

Supplier: Thermo Scientific
Warning

Synonyms: Methyl 6-(trifluoromethyl)nicotinate

H33836.MD H33836.03
H33836.MDEA 0 EUR
H33836.MD H33836.03
Methyl-2-(trifluoromethyl)pyridine-5-carboxylate 97%
Methyl-2-(trifluoromethyl)pyridine-5-carboxylate

Purity: 97% CAS No.: 221313-10-6 Molecular Formula: C8H6F3NO2 Formula Weight: 205.14 Melting Point: 54-56° MDL No.: MFCD03001346

Formula: C₈H₆F₃NO₂
MW: 205,13 g/mol
Melting Pt: 54…56 °C
Storage Temperature: Ambient
MDL Number: MFCD03001346
CAS Number: 221313-10-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR