3-(4-Fluoro-3-methoxyphenyl)propionic acid 96%
Supplier: Thermo Scientific
|
Warning
|
Synonyms:
3-(4-Fluoro-3-methoxyphenyl)propanoic acid
H33894.03
H33894.MD
H33894.03EA
187
EUR
H33894.03
H33894.MD
3-(4-Fluoro-3-methoxyphenyl)propionic acid 96%
3-(4-Fluoro-3-methoxyphenyl)propionic acid
Purity: 96%
CAS No.: 864960-96-3
Molecular Formula: C10H11FO3
Formula Weight: 198.19
Melting Point: 90-94°
MDL No.: MFCD11655589
Formula:
C₁₀H₁₁FO₃ MW: 198,19 g/mol Melting Pt: 90…94 °C Storage Temperature: Ambient |
MDL Number:
MFCD11655589 CAS Number: 864960-96-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...