3-(5-Bromo-2-methoxyphenyl)propionic acid 96%
Supplier: Thermo Scientific
|
Warning
|
Synonyms:
3-(5-Bromo-2-methoxyphenyl)propanoic acid
H33961.03
H33961.06
H33961.03EA
76.8
EUR
H33961.03
H33961.06
3-(5-Bromo-2-methoxyphenyl)propionic acid 96%
3-(5-Bromo-2-methoxyphenyl)propionic acid
Purity: 96%
CAS No.: 82547-30-6
Molecular Formula: C10H11BrO3
Formula Weight: 259.10
Melting Point: 114-116°
MDL No.: MFCD09258907
Formula:
C₁₀H₁₁BrO₃ MW: 259,10 g/mol Melting Pt: 114…116 °C Storage Temperature: Ambient |
MDL Number:
MFCD09258907 CAS Number: 82547-30-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...