Ethyl-2-(pyridin-3-yloxy)acetate 98%

Supplier: Thermo Scientific
Warning

H33981.03 H33981.09
H33981.03EA 0 EUR
H33981.03 H33981.09
Ethyl-2-(pyridin-3-yloxy)acetate 98%
Ethyl-2-(pyridin-3-yloxy)acetate

Purity: 98% CAS No.: 18342-98-8 Molecular Formula: C9H11NO3 Formula Weight: 181.19 Refractive Index: 1.506 MDL No.: MFCD00093962

Formula: C₉H₁₁NO₃
MW: 181,19 g/mol
Density: 1,157
Storage Temperature: Ambient
MDL Number: MFCD00093962
CAS Number: 18342-98-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR