Diethyl-5-[(chloroacetyl)amino]-3-methylthiophene-2,4-dicarboxylate 96%

Supplier: Thermo Fisher Scientific
Warning

H34228.MD H34228.03
H34228.MDEA 0 EUR
H34228.MD H34228.03
Diethyl-5-[(chloroacetyl)amino]-3-methylthiophene-2,4-dicarboxylate 96%
Diethyl-5-[(chloroacetyl)amino]-3-methylthiophene-2,4-dicarboxylate

Purity: 96% CAS No.: 146381-87-5 Molecular Formula: C13H16ClNO5S Formula Weight: 333.79 Melting Point: 164-169° MDL No.: MFCD00414318

Formula: C₁₃H₁₆ClNO₅S
Melting Pt: 164…169 °C
Storage Temperature: Ambient
MDL Number: MFCD00414318
CAS Number: 146381-87-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR