3-Allyl-5,6-dimethyl-2-thioxo-2,3-dihydrothieno[2,3-d]pyrimidin-4(1H)-one 96%
Supplier: Thermo Scientific
|
Warning
|
H34437.MD
H34437.03
H34437.MDEA
0
EUR
H34437.MD
H34437.03
3-Allyl-5,6-dimethyl-2-thioxo-2,3-dihydrothieno[2,3-d]pyrimidin-4(1H)-one 96%
3-Allyl-5,6-dimethyl-2-thioxo-2,3-dihydrothieno[2,3-d]pyrimidin-4(1H)-one
Purity: 96%
CAS No.: 51486-16-9
Molecular Formula: C11H12N2OS2
Formula Weight: 252.36
Melting Point: 202-203°
MDL No.: MFCD01911592
Formula:
C₁₁H₁₂N₂OS₂ MW: 252,36 g/mol Melting Pt: 202…203 °C Storage Temperature: Ambient |
MDL Number:
MFCD01911592 CAS Number: 51486-16-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...