N-(3-Chloro-5-trifluoromethyl-2-pyridyl)thiourea 97%
Supplier: Thermo Scientific
|
Danger
|
H34440.MD
H34440.03
H34440.06
H34440.MDEA
39
EUR
H34440.MD
H34440.03
H34440.06
N-(3-Chloro-5-trifluoromethyl-2-pyridyl)thiourea 97%
N-(3-Chloro-5-trifluoromethyl-2-pyridyl)thiourea
Purity: 97%
Molecular Formula: C7H5ClF3N3S
Formula Weight: 255.64
Melting Point: 189-190°
MDL No.: MFCD00141984
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...