1-Ethynylpyrene 98
Supplier: Thermo Fisher Scientific
H37327.MD
H37327.03
H37327.06
H37327.MDEA
34.2
EUR
H37327.MD
H37327.03
H37327.06
1-Ethynylpyrene 98
1-Ethynylpyrene
Purity: 98+%
CAS No.: 34993-56-1
Molecular Formula: C18H10
Physical Form: Powder
Appearance: Yellow
Melting Point: 113-116°
Beilstein Reference: 2365008
MDL No.: MFCD02093933
Formula:
C₁₈H₁₀ MW: 226.28 g/mol Melting Pt: 113…116 °C Storage Temperature: Ambient |
MDL Number:
MFCD02093933 CAS Number: 34993-56-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...