6-Bromo-8-iodoimidazo[1,2-a]pyridine 95%
Supplier: Thermo Scientific
|
Warning
|
H37542.03
H37542.06
H37542.03EA
0
EUR
H37542.03
H37542.06
6-Bromo-8-iodoimidazo[1,2-a]pyridine 95%
6-Bromo-8-iodoimidazo[1,2-a]pyridine
Purity: 95%
Molecular Formula: C5H4BrIN2
Formula Weight: 322.92
Physical Form: Powder
Sensitivity: Light Sensitive
Appearance: Yellow
Melting Point: 147-150°
MDL No.: MFCD21609484
Formula:
C₇H₄BrIN₂ Melting Pt: 147…150 °C Storage Temperature: Ambient |
MDL Number:
MFCD21609484 CAS Number: 1364917-14-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...