2-Amino-3-pyridinemethanol 98%

Supplier: Thermo Scientific
Danger

Synonyms: (2-Amino-pyridin-3-yl)-methanol, 3-(Hydroxymethyl)-2-pyridinamine, 2-Amino-3-pyridinemethanol, 2-Amino-3-hydroxymethylpyridine, 2-Amino-3-pyridylmethanol, 2-Amino-3-(hydroxymethyl)pyridine, 2-Aminopyridine-3-methanol

H50010.03 H50010.06
H50010.03EA 0 EUR
H50010.03 H50010.06
2-Amino-3-pyridinemethanol 98%
2-Amino-3-pyridinemethanol

Purity: 98% CAS No.: 23612-57-9 Molecular Formula: C6H8N2O Formula Weight: 124.14 MDL No.: MFCD05663510

Formula: C₆H₈N₂O
MW: 124,14 g/mol
Storage Temperature: Ambient
MDL Number: MFCD05663510
CAS Number: 23612-57-9
UN: 2811
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR