(2-Chloro-4-iodo-3-pyridinyl)methanol 97%

Supplier: Thermo Scientific
Warning

H50052.03 H50052.MD
H50052.03EA 210 EUR
H50052.03 H50052.MD
(2-Chloro-4-iodo-3-pyridinyl)methanol 97%
(2-Chloro-4-iodo-3-pyridinyl)methanol

Purity: 97% CAS No.: 884494-44-4 Molecular Formula: C6H5ClINO Formula Weight: 269.47 MDL No.: MFCD03095296

Formula: C₆H₅ClINO
MW: 269,47 g/mol
Storage Temperature: Ambient
MDL Number: MFCD03095296
CAS Number: 884494-44-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR