4,6-Dimethylisoxazolo[5,4-b]pyridin-3-ylamine 95%
Supplier: Thermo Scientific
|
Warning
|
H50122.03
H50122.MD
H50122.03EA
0
EUR
H50122.03
H50122.MD
4,6-Dimethylisoxazolo[5,4-b]pyridin-3-ylamine 95%
4,6-Dimethylisoxazolo[5,4-b]pyridin-3-ylamine
Purity: 95%
CAS No.: 916792-12-6
Molecular Formula: C8H9N3O
Formula Weight: 163.18
MDL No.: MFCD08060940
Formula:
C₈H₉N₃O Storage Temperature: Ambient |
MDL Number:
MFCD08060940 CAS Number: 916792-12-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...