2-Cyclopentyloxypyridine-5-carboxaldehyde 98%
Supplier: Thermo Scientific
|
Warning
|
H50128.03
H50128.06
H50128.03EA
186
EUR
H50128.03
H50128.06
2-Cyclopentyloxypyridine-5-carboxaldehyde 98%
2-Cyclopentyloxypyridine-5-carboxaldehyde
Purity: 98%
CAS No.: 916792-14-8
Molecular Formula: C11H13NO2
Formula Weight: 191.23
MDL No.: MFCD08060943
Formula:
C₁₁H₁₃NO₂ MW: 191,23 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD08060943 CAS Number: 916792-14-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...