Ethyl 2-chloro-5-pyridylglyoxylate 97%

Supplier: Thermo Fisher Scientific

H50157.03 H50157.06
H50157.03EA 97.5 EUR
H50157.03 H50157.06
Ethyl 2-chloro-5-pyridylglyoxylate 97%
Ethyl-2-chloro-5-pyridylglyoxylate

Purity: 97% CAS No.: 902837-55-2 Molecular Formula: C9H8ClNO3 Formula Weight: 213.62 MDL No.: MFCD07781188

Formula: C₉H₈ClNO₃
Storage Temperature: Ambient
MDL Number: MFCD07781188
CAS Number: 902837-55-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR