4-(Trifluoromethoxy)phenoxyacetic acid 95%

Supplier: Thermo Fisher Scientific
Warning

Synonyms: 2-(4-(Trifluoromethoxy)phenoxy)acetic acid

H50218.03 H50218.06
H50218.03EA 0 EUR
H50218.03 H50218.06
4-(Trifluoromethoxy)phenoxyacetic acid 95%
4-(Trifluoromethoxy)phenoxyacetic acid

Purity: 95% CAS No.: 72220-50-9 Molecular Formula: C9H7F3O4 Formula Weight: 236.15 MDL No.: MFCD00052352

Formula: C₉H₇F₃O₄
Storage Temperature: Ambient
MDL Number: MFCD00052352
CAS Number: 72220-50-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR