3-Benzyloxy-2-hydroxy-1-propylamine 99%

Supplier: Thermo Fisher Scientific

H50248.03 H50248.MD
H50248.03EA 0 EUR
H50248.03 H50248.MD
3-Benzyloxy-2-hydroxy-1-propylamine 99%
3-Benzyloxy-2-hydroxy-1-propylamine

Purity: 99% CAS No.: 90503-15-4 Molecular Formula: C10H15NO2 Formula Weight: 181.24 MDL No.: MFCD00183222

Formula: C₁₀H₁₅NO₂
Storage Temperature: Ambient
MDL Number: MFCD00183222
CAS Number: 90503-15-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR