2-(2-Methoxyphenoxy)ethylamine 97%

Supplier: Thermo Scientific
Danger

Synonyms: 1-(2-Aminoethoxy)-2-methoxybenzene

H50251.MD
H50251.MDEA 93.4 EUR
H50251.MD
2-(2-Methoxyphenoxy)ethylamine 97%
2-(2-Methoxyphenoxy)ethylamine

Purity: 97% CAS No.: 1836-62-0 Molecular Formula: C9H13NO2 Formula Weight: 167.21 Odour: Amine-like Boiling Point: 98°/0.4mm UN No.: UN2735 MDL No.: MFCD00235185

Formula: C₉H₁₃NO₂
MW: 167,21 g/mol
Boiling Pt: 98 °C (0,4 mmHg)
Density: 1,11
Storage Temperature: Ambient
MDL Number: MFCD00235185
CAS Number: 1836-62-0
UN: 2735
ADR: 8,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR