3-Oxo-2,3-dihydro-4H-1,4-benzoxazine-4-acetic acid 98%

Supplier: Thermo Scientific
Warning

Synonyms: 2-(3-Oxo-2H-benzo[b][1,4]oxazin-4(3H)-yl)acetic acid

H50275.03 H50275.MD
H50275.03EA 516 EUR
H50275.03 H50275.MD
3-Oxo-2,3-dihydro-4H-1,4-benzoxazine-4-acetic acid 98%
3-Oxo-2,3-dihydro-4H-1,4-benzoxazine-4-acetic acid

Purity: 98% CAS No.: 26494-55-3 Molecular Formula: C10H9NO4 Formula Weight: 207.19 MDL No.: MFCD00269587

Formula: C₁₀H₉NO₄
MW: 207,19 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00269587
CAS Number: 26494-55-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR