6-Chloroimidazo[1,2-a]pyridine-2-carboxylic acid 99%
Supplier: Thermo Scientific
|
Warning
|
H50313.MD
H50313.MDEA
90.5
EUR
H50313.MD
6-Chloroimidazo[1,2-a]pyridine-2-carboxylic acid 99%
6-Chloroimidazo[1,2-a]pyridine-2-carboxylic acid
Purity: 99%
CAS No.: 182181-19-7
Molecular Formula: C8H5ClN2O2
Formula Weight: 196.59
Melting Point: 246-247°
MDL No.: MFCD02186559
Formula:
C₈H₅ClN₂O₂ MW: 196,59 g/mol Melting Pt: 246…247 °C Storage Temperature: Ambient |
MDL Number:
MFCD02186559 CAS Number: 182181-19-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...