3-(4-Chlorosulphonylphenyl)propionic acid 99%
Supplier: Thermo Scientific
|
Danger
|
Synonyms:
4-(Chlorosulphonyl)dihydrocinnamic acid
H50384.03
H50384.06
H50384.03EA
178
EUR
H50384.03
H50384.06
3-(4-Chlorosulphonylphenyl)propionic acid 99%
3-(4-Chlorosulphonylphenyl)propionic acid
Purity: 99%
CAS No.: 63545-54-0
Molecular Formula: C9H9ClO4S
Formula Weight: 248.69
UN No.: UN3261
MDL No.: MFCD02179510
Formula:
C₉H₉ClO₄S MW: 248,69 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD02179510 CAS Number: 63545-54-0 ADR: 8,II |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...