2-(2-Methyl-4-thiazolyl)phenol 97%
Supplier: Thermo Scientific
|
Warning
|
Synonyms:
2-(2-Methylthiazol-4-yl)phenol
H50431.03
H50431.MD
H50431.03EA
0
EUR
H50431.03
H50431.MD
2-(2-Methyl-4-thiazolyl)phenol 97%
2-(2-Methyl-4-thiazolyl)phenol
Purity: 97%
CAS No.: 160241-65-6
Molecular Formula: C10H9NOS
Formula Weight: 191.25
Melting Point: 61-64°
MDL No.: MFCD00665949
Formula:
C₁₀H₉NOS MW: 191,25 g/mol Melting Pt: 61…64 °C Storage Temperature: Ambient |
MDL Number:
MFCD00665949 CAS Number: 160241-65-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...