Biphenyl-3-carboxaldehyde 95%

Supplier: Thermo Scientific
Warning

Synonyms: 3-Phenylbenzaldehyde, [1,1'-Biphenyl]-3-carbaldehyde

H50453.03
H50453.03EA 233 EUR
H50453.03
Biphenyl-3-carboxaldehyde 95%
Biphenyl-3-carboxaldehyde

Purity: 95% CAS No.: 1204-60-0 Molecular Formula: C13H10O Formula Weight: 182.22 MDL No.: MFCD01740432

Formula: C₁₃H₁₀O
MW: 182,22 g/mol
Storage Temperature: Ambient
MDL Number: MFCD01740432
CAS Number: 1204-60-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR