2,4-Dichloro-5-thiazolecarboxaldehyde 96%
Supplier: Thermo Scientific
|
Warning
|
Synonyms:
2,4-Dichlorothiazole-5-carbaldehyde
H50456.03
H50456.MD
H50456.03EA
340
EUR
H50456.03
H50456.MD
2,4-Dichloro-5-thiazolecarboxaldehyde 96%
2,4-Dichloro-5-thiazolecarboxaldehyde
Purity: 96%
CAS No.: 92972-48-0
Molecular Formula: C4HCl2NOS
Formula Weight: 182.03
Melting Point: 48-49°
MDL No.: MFCD00793013
Formula:
C₄HCl₂NOS MW: 182,03 g/mol Melting Pt: 48…49 °C Storage Temperature: Ambient |
MDL Number:
MFCD00793013 CAS Number: 92972-48-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...