2-Morpholino-1,3-thiazole-5-carbaldehyde 97%
Supplier: Thermo Scientific
|
Warning
|
H50463.03
H50463.MD
H50463.03EA
314
EUR
H50463.03
H50463.MD
2-Morpholino-1,3-thiazole-5-carbaldehyde 97%
2-Morpholino-1,3-thiazole-5-carbaldehyde
Purity: 97%
CAS No.: 1011-41-2
Molecular Formula: C8H10N2O2S
Formula Weight: 198.25
MDL No.: MFCD01568825
Formula:
C₈H₁₀N₂O₂S MW: 198,25 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD01568825 CAS Number: 1011-41-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...