2-Chloro-3-cyano-6-(4-methoxyphenyl)pyridine 98%

Supplier: Thermo Scientific
Warning

H50520.03
H50520.03EA 291 EUR
H50520.03
2-Chloro-3-cyano-6-(4-methoxyphenyl)pyridine 98%
2-Chloro-3-cyano-6-(4-methoxyphenyl)pyridine

Purity: 98% CAS No.: 306977-91-3 Molecular Formula: C13H9ClN2O Formula Weight: 244.68 Melting Point: 148° dec. MDL No.: MFCD00215274

Formula: C₁₃H₉ClN₂O
MW: 244,68 g/mol
Melting Pt: 148 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD00215274
CAS Number: 306977-91-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR