5-Amino-2-(2,4-dichlorophenoxy)pyridine 99%

Supplier: Thermo Scientific

Synonyms: 6-(2,4-Dichlorophenoxy)pyridin-3-amine

H50683.03 H50683.MD
H50683.03EA 307 EUR
H50683.03 H50683.MD
5-Amino-2-(2,4-dichlorophenoxy)pyridine 99%
5-Amino-2-(2,4-dichlorophenoxy)pyridine

Purity: 99% CAS No.: 85686-48-2 Molecular Formula: C11H8Cl2N2O Formula Weight: 255.11 MDL No.: MFCD00052643

Formula: C₁₁H₈Cl₂N₂O
MW: 255,11 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00052643
CAS Number: 85686-48-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR