1-(Dimethylaminomethyl)cyclopentanecarboxaldehyde 97%

Supplier: Thermo Scientific
Warning

H50751.03 H50751.MD
H50751.03EA 0 EUR
H50751.03 H50751.MD
1-(Dimethylaminomethyl)cyclopentanecarboxaldehyde 97%
1-(Dimethylaminomethyl)cyclopentanecarboxaldehyde

Purity: 97% CAS No.: 39943-37-8 Molecular Formula: C9H17NO Formula Weight: 155.24 MDL No.: MFCD08064242

Formula: C₉H₁₇NO
MW: 155,24 g/mol
Storage Temperature: Ambient
MDL Number: MFCD08064242
CAS Number: 39943-37-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR