1-(3-Methoxyphenyl)-2-(methylamino)ethanol 98%

Supplier: Thermo Scientific
Warning

H50758.MD
H50758.MDEA 93.9 EUR
H50758.MD
1-(3-Methoxyphenyl)-2-(methylamino)ethanol 98%
1-(3-Methoxyphenyl)-2-(methylamino)ethanol

Purity: 98% CAS No.: 92188-49-3 Molecular Formula: C10H15NO2 Formula Weight: 181.24 MDL No.: MFCD08060971

Formula: C₁₀H₁₅NO₂
MW: 181,24 g/mol
Storage Temperature: Ambient
MDL Number: MFCD08060971
CAS Number: 92188-49-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR