3-(Benzyloxy)pyrrolidine 96%

Supplier: Thermo Fisher Scientific
Warning

H50769.03 H50769.MD
H50769.03EA 266 EUR
H50769.03 H50769.MD
3-(Benzyloxy)pyrrolidine 96%
3-(Benzyloxy)pyrrolidine

Purity: 96% CAS No.: 420137-14-0 Molecular Formula: C11H15NO Formula Weight: 177.25 MDL No.: MFCD08060975

Formula: C₁₁H₁₅NO
Storage Temperature: Ambient
MDL Number: MFCD08060975
CAS Number: 420137-14-0

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR