tert-Butyl-3-oxoazetidine-1-carboxylate 95%
Supplier: Thermo Scientific
Synonyms:
N-Boc-3-azetidinone, Tert-butyl 3-oxoazetidine-1-carboxylate, tert-Butyl 3-oxoazetidine-1-carboxylate, 1-BOC-3-Azetidinone
H50826.03
H50826.MD
H50826.03EA
129
EUR
H50826.03
H50826.MD
tert-Butyl-3-oxoazetidine-1-carboxylate 95%
tert-Butyl-3-oxoazetidine-1-carboxylate
Purity: 95%
CAS No.: 398489-26-4
Molecular Formula: C8H13NO3
Formula Weight: 171.20
Melting Point: 49-52°
MDL No.: MFCD01861741
Formula:
C₈H₁₃NO₃ MW: 171,2 g/mol Melting Pt: 49…52 °C Storage Temperature: Ambient |
MDL Number:
MFCD01861741 CAS Number: 398489-26-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...