(2,6-Dimethoxy-phenoxy)-acetic acid ≥98%
Supplier: Thermo Fisher Scientific
|
Warning
|
Synonyms:
2-(2,6-Dimethoxyphenoxy)acetic acid
H50894.MD
H50894.06
H50894.MDEA
0
EUR
H50894.MD
H50894.06
(2,6-Dimethoxy-phenoxy)-acetic acid ≥98%
(2,6-Dimethoxyphenoxy)acetic acid
Purity: 98+%
CAS No.: 95110-10-4
Molecular Formula: C10H12O5
Formula Weight: 212.20
Melting Point: 79-82°
MDL No.: MFCD06823823
Formula:
C₁₀H₁₂O₅ Melting Pt: 79…82 °C Storage Temperature: Ambient |
MDL Number:
MFCD06823823 CAS Number: 95110-10-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...