1-(6-Ethoxy-2-pyridyl)piperazine 97%

Supplier: Thermo Scientific
Warning

H50905.03 H50905.MD
H50905.03EA 0 EUR
H50905.03 H50905.MD
1-(6-Ethoxy-2-pyridyl)piperazine 97%
1-(6-Ethoxy-2-pyridyl)piperazine

Purity: 97% CAS No.: 108122-25-4 Molecular Formula: C11H17N3O Formula Weight: 207.28 MDL No.: MFCD08061051

Formula: C₁₁H₁₇N₃O
MW: 207,28 g/mol
Storage Temperature: Ambient
MDL Number: MFCD08061051
CAS Number: 108122-25-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR