4-Propoxypiperidine 95%

Supplier: Thermo Fisher Scientific
Warning

H50976.03 H50976.MD
H50976.03EA 323 EUR
H50976.03 H50976.MD
4-Propoxypiperidine 95%
4-Propoxypiperidine

Purity: 95% CAS No.: 88536-11-2 Molecular Formula: C8H17NO Formula Weight: 143.23 MDL No.: MFCD06248727

Formula: C₈H₁₇NO
Storage Temperature: Ambient
MDL Number: MFCD06248727
CAS Number: 88536-11-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR