Methyl-3-[4-(aminomethyl)phenyl]propionate hydrochloride 97%

Supplier: Thermo Scientific
Warning

H51031.MD H51031.03
H51031.MDEA 0 EUR
H51031.MD H51031.03
Methyl-3-[4-(aminomethyl)phenyl]propionate hydrochloride 97%
Methyl-3-[4-(aminomethyl)phenyl]propionate hydrochloride

CAS No.: 103565-40-8 Molecular Formula: C11H15NO2.HCl Formula Weight: 229.71 Beilstein Reference: 14326402 MDL No.: MFCD11113035

Formula: C₁₁H₁₅NO₂·HCl
MW: 229,71 g/mol
Storage Temperature: Ambient
MDL Number: MFCD11113035
CAS Number: 103565-40-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR