1,4-Phenylenediacryloyl chloride, tech.

Supplier: Thermo Scientific
Danger

Synonyms: 3,3'-Benzene-1,4-diylbisprop-2-enoyl chloride, 3,3'-(1,4-Phenylene)diacryloyl chloride

H51055.MD H51055.03
H51055.MDEA 0 EUR
H51055.MD H51055.03
1,4-Phenylenediacryloyl chloride, tech.
1,4-Phenylenediacryloyl chloride

CAS No.: 35288-49-4 Molecular Formula: C12H8Cl2O2 Formula Weight: 255.10 UN No.: UN1759 MDL No.: MFCD01109676

Formula: C₁₂H₈Cl₂O₂
MW: 255,10 g/mol
Storage Temperature: Ambient
MDL Number: MFCD01109676
CAS Number: 35288-49-4
UN: 3261
ADR: 8,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR