2-Butenylmagnesium chloride 0.5 M in MeTHF (2-methyltetrahydrofuran)

Supplier: Thermo Fisher Scientific
Danger

H51171.AE H51171.AP
H51171.AEEA 240 EUR
H51171.AE H51171.AP
2-Butenylmagnesium chloride 0.5 M in MeTHF (2-methyltetrahydrofuran)
2-Butenylmagnesium chloride

CAS No.: 22649-70-3 Molecular Formula: C4H7ClMg Formula Weight: 114.86 UN No.: UN3399 MDL No.: MFCD01321226

Formula: C₄H₇ClMg
MW: 114.86 g/mol
Storage Temperature: Ambient
MDL Number: MFCD01321226
CAS Number: 6088-88-6
UN: 3399
ADR: 4.3 II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR